Learn More
Medchemexpress LLC PF-06424439 methanesulfonate | 1469284-79-4 | 99.9% | 536.05 g/mol | C23H30ClN7O4S | 100 MG
SDP
Catalog No. 5000243668
Encompass_Preferred
PF-06424439 methanesulfonate is the methanesulfonate salt form of a potent, selective small-molecule inhibitor of diacylglycerol acyltransferase 2 (DGAT2). It is supplied as a high-purity solid for preclinical research to study triglyceride synthesis and lipid metabolism in biochemical, cellular, and animal models.
  • Potent, selective DGAT2 inhibitor (IC50 14 nM).
  • High purity suitable for research applications.
  • Methanesulfonate salt to aid solubility and handling.
  • Suitable for in vitro and in vivo preclinical studies.
  • Supplied as a 100 mg pack for controlled dosing.
Catalog No. 50-002-43668 Supplier Medchemexpress LLC Supplier No. HY108341A100MG
PF-06424439 methanesulfonate is the methanesulfonate salt form of a potent, selective small-molecule inhibitor of diacylglycerol acyltransferase 2 (DGAT2). It is supplied as a high-purity solid for preclinical research to study triglyceride synthesis and lipid metabolism in biochemical, cellular, and animal models.
  • Potent, selective DGAT2 inhibitor (IC50 14 nM).
  • High purity suitable for research applications.
  • Methanesulfonate salt to aid solubility and handling.
  • Suitable for in vitro and in vivo preclinical studies.
  • Supplied as a 100 mg pack for controlled dosing.