Learn More
Medchemexpress LLC PF-06424439 methanesulfonate | 1469284-79-4 | 99.9% | 536.05 g/mol | C23H30ClN7O4S | 25 MG
SDP
Catalog No. 5000244326
Encompass_Preferred
PF-06424439 methanesulfonate is a research-grade small-molecule inhibitor of diacylglycerol acyltransferase 2 (DGAT2). It is a potent, selective imidazopyridine that inhibits DGAT2 noncompetitively (IC50 = 14 nM) and exhibits slowly reversible, time-dependent inhibition. Supplied as a methanesulfonate salt for laboratory research.
  • Potent, selective DGAT2 inhibitor (IC50 = 14 nM).
  • Noncompetitive, slowly reversible, time-dependent mode of inhibition.
  • High purity (99.9%).
  • Solid, white to yellow physical form.
  • High solubility in DMSO and water for in vitro use (DMSO 250 mg/mL; H2O 50 mg/mL).
  • Available in small pack sizes suitable for preclinical research.
Catalog No. 50-002-44326 Supplier Medchemexpress LLC Supplier No. HY108341A25MG
PF-06424439 methanesulfonate is a research-grade small-molecule inhibitor of diacylglycerol acyltransferase 2 (DGAT2). It is a potent, selective imidazopyridine that inhibits DGAT2 noncompetitively (IC50 = 14 nM) and exhibits slowly reversible, time-dependent inhibition. Supplied as a methanesulfonate salt for laboratory research.
  • Potent, selective DGAT2 inhibitor (IC50 = 14 nM).
  • Noncompetitive, slowly reversible, time-dependent mode of inhibition.
  • High purity (99.9%).
  • Solid, white to yellow physical form.
  • High solubility in DMSO and water for in vitro use (DMSO 250 mg/mL; H2O 50 mg/mL).
  • Available in small pack sizes suitable for preclinical research.