Learn More
Medchemexpress LLC HY-120088A 10mg Medchemexpress, PF-06446846 (hydrochloride) CAS:1632250-50-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120088A10MG
Encompass_Preferred
Medchemexpress, HY-120088A 10mg PF-06446846 (HCl) CAS:1632250-50-0 PF-06446846 hydrochloride is an orally active and highly selective inhibitor of translation of Proprotein convertase subtilisin/kexin type 9 (PCSK9). PF-06446846 hydrochloride inhibits PCSK9 by inducing the ribosome to stall around codon 34. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9572