Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pf-06446846 hydrochloride | 1632250-50-0 | 99.4% | 10 MM 1 ML
SDP

Catalog No. 5000231863
Encompass_Preferred

Pf-06446846 hydrochloride is the hydrochloride salt of a small-molecule inhibitor that selectively inhibits translation of PCSK9 by inducing ribosomal stalling; supplied as a 10 mM solution in DMSO for research use. It reduces PCSK9 secretion in Huh7 cells (IC50 ≈ 0.3 μM).

  • Hydrochloride salt form, soluble in DMSO and water.
  • Targets PCSK9 and inhibits its translation via ribosomal stalling.
  • Demonstrated activity in cell assays (Huh7 cells; IC50 ~0.3 μM).
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • Molecular weight 470.35; formula C22H21Cl2N7O.
  • Storage: solid at 4°C; in solvent, -80°C (up to 2 years) or -20°C (up to 1 year).

Catalog No. 50-002-31863 Supplier Medchemexpress LLC Supplier No. HY120088A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Pf-06446846 hydrochloride is the hydrochloride salt of a small-molecule inhibitor that selectively inhibits translation of PCSK9 by inducing ribosomal stalling; supplied as a 10 mM solution in DMSO for research use. It reduces PCSK9 secretion in Huh7 cells (IC50 ≈ 0.3 μM).

  • Hydrochloride salt form, soluble in DMSO and water.
  • Targets PCSK9 and inhibits its translation via ribosomal stalling.
  • Demonstrated activity in cell assays (Huh7 cells; IC50 ~0.3 μM).
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • Molecular weight 470.35; formula C22H21Cl2N7O.
  • Storage: solid at 4°C; in solvent, -80°C (up to 2 years) or -20°C (up to 1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.