Learn More
Medchemexpress LLC PF-06446846 (hydrochloride) | 1632250-50-0 | 99.4% | 470.35 | 5 MG
Supplier: Medchemexpress LLC 5MGHY120088A
PF-06446846 hydrochloride is an orally active and highly selective inhibitor of translation of Proprotein convertase subtilisin/kexin type 9 (PCSK9). It inhibits PCSK9 by inducing the ribosome to stall around codon 34. For research use only.
- High purity (99.4%)
- Solid appearance
- Light yellow to yellow color