Learn More
Selleck Chemical LLC PF-06446846 hydrochloride
SDP
Catalog No. 503136017
Encompass_Preferred
PF-06446846 (PF846) Hydrochloride is an orally bioavailable and specific inhibitor of PCSK9 with an IC50 of 0 3 M in Huh7 cells
Catalog No. 50-313-6017 Supplier Selleck Chemical LLC Supplier No. E011625MG
PF-06446846 (PF846) Hydrochloride is an orally bioavailable and specific inhibitor of PCSK9 with an IC50 of 0 3 M in Huh7 cells