Learn More
Medchemexpress LLC HY-12477 5mg Medchemexpress, PF-06447475 CAS:1527473-33-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY124775MG
Encompass_Preferred
Medchemexpress, HY-12477 5mg PF-06447475 CAS:1527473-33-1 PF-06447475 is a highly potent, selective and brain penetrant LRRK2 inhibitor with an IC50 of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8884