Learn More
Medchemexpress LLC PF-06456384 trihydrochloride | 1834610-75-1 | ≥98.0% | 829.18 g·mol⁻¹ | C35H32F3N7O3S2 · 3HCl | 25 MG
SDP
Catalog No. 5000308073
Encompass_Preferred
The trihydrochloride salt is a small-molecule research compound described as a highly potent and selective inhibitor of the NaV1.7 voltage-gated sodium channel; it has reported sub-nanomolar activity and is used in preclinical pain model research.
  • Highly potent and selective NaV1.7 inhibition (reported IC50 ≈ 0.01 nM).
  • Used in preclinical pain models, including the formalin test.
  • Supplied as a trihydrochloride salt in white to beige powder form.
  • High purity (≥98% by HPLC reported by distributors).
  • Solubility: readily soluble in DMSO (~125 mg/mL); limited aqueous solubility (~2 mg/mL reported).
Catalog No. 50-003-08073 Supplier Medchemexpress LLC Supplier No. HY118952A25MG
The trihydrochloride salt is a small-molecule research compound described as a highly potent and selective inhibitor of the NaV1.7 voltage-gated sodium channel; it has reported sub-nanomolar activity and is used in preclinical pain model research.
  • Highly potent and selective NaV1.7 inhibition (reported IC50 ≈ 0.01 nM).
  • Used in preclinical pain models, including the formalin test.
  • Supplied as a trihydrochloride salt in white to beige powder form.
  • High purity (≥98% by HPLC reported by distributors).
  • Solubility: readily soluble in DMSO (~125 mg/mL); limited aqueous solubility (~2 mg/mL reported).