Learn More
Medchemexpress LLC PF-06456384 trihydrochloride | 1834610-75-1 | 99.4% | 829.18 g/mol | C35H35Cl3F3N7O3S2 | 100 MG
SDP
Catalog No. 5000230019
Encompass_Preferred
PF-06456384 trihydrochloride is a potent, selective small-molecule inhibitor of the voltage-gated sodium channel NaV1.7 for laboratory research use. Provided as the trihydrochloride salt, it appears as a white to off-white solid with high listed purity and is used in preclinical pain-model studies. Available in solid and solution formats for flexible assay setup.
  • Potent NaV1.7 inhibition (IC50 = 0.01 nM), enabling high-sensitivity pharmacology studies.
  • Trihydrochloride salt form for improved solubility and handling.
  • High listed purity for reproducible experimental results.
  • Available in multiple pack sizes and as a 10 mM DMSO solution for assay flexibility.
  • Intended for laboratory research use only; not for human or veterinary use.
Catalog No. 50-002-30019 Supplier Medchemexpress LLC Supplier No. HY118952A100MG
PF-06456384 trihydrochloride is a potent, selective small-molecule inhibitor of the voltage-gated sodium channel NaV1.7 for laboratory research use. Provided as the trihydrochloride salt, it appears as a white to off-white solid with high listed purity and is used in preclinical pain-model studies. Available in solid and solution formats for flexible assay setup.
  • Potent NaV1.7 inhibition (IC50 = 0.01 nM), enabling high-sensitivity pharmacology studies.
  • Trihydrochloride salt form for improved solubility and handling.
  • High listed purity for reproducible experimental results.
  • Available in multiple pack sizes and as a 10 mM DMSO solution for assay flexibility.
  • Intended for laboratory research use only; not for human or veterinary use.