Learn More
Medchemexpress LLC Pf-06456384 trihydrochloride | 1834610-75-1 | 99.4% | 829.18 g/mol | C35H35Cl3F3N7O3S2 | 5 MG
SDP
Catalog No. 5000226417
Encompass_Preferred
PF-06456384 trihydrochloride is a research-grade small-molecule trihydrochloride salt and a highly potent, selective NaV1.7 inhibitor (reported IC50 = 0.01 nM) provided for preclinical pharmacology and pain-model studies.
  • High potency and selectivity toward NaV1.7 for target validation and pharmacology studies.
  • Trihydrochloride salt form improves compound handling and solubility for assays.
  • High purity (reported ~99.38%) suitable for research applications.
  • Molecular weight 829.18 g/mol; white to off-white solid.
  • Supplied in small research quantities for assay development and in vivo pilot studies.
Catalog No. 50-002-26417 Supplier Medchemexpress LLC Supplier No. HY118952A5MG
PF-06456384 trihydrochloride is a research-grade small-molecule trihydrochloride salt and a highly potent, selective NaV1.7 inhibitor (reported IC50 = 0.01 nM) provided for preclinical pharmacology and pain-model studies.
  • High potency and selectivity toward NaV1.7 for target validation and pharmacology studies.
  • Trihydrochloride salt form improves compound handling and solubility for assays.
  • High purity (reported ~99.38%) suitable for research applications.
  • Molecular weight 829.18 g/mol; white to off-white solid.
  • Supplied in small research quantities for assay development and in vivo pilot studies.