Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-06456384 trihydrochloride | 1834610-75-1 | 99.4% | 829.18 g/mol | C35H35Cl3F3N7O3S2 | 50 MG
SDP

Catalog No. 5000228884
Encompass_Preferred

PF-06456384 trihydrochloride is the trihydrochloride salt of a highly potent and selective NaV1.7 inhibitor developed for preclinical pharmacology and pain model research. The compound has been profiled in vitro and in vivo and is supplied as a high-purity research sample.

  • Salt form: trihydrochloride.
  • Target: voltage-gated sodium channel NaV1.7.
  • Potency: IC50 = 0.01 nM against NaV1.7.
  • Purity: 99.38% by HPLC (supplier specification).
  • Molecular weight: 829.18 g/mol (trihydrochloride salt).
  • Molecular formula: C35H35Cl3F3N7O3S2.
  • Intended use: preclinical pharmacology and formalin pain model research.
  • Supplied quantity: 50 mg research sample.

Catalog No. 50-002-28884 Supplier Medchemexpress LLC Supplier No. HY118952A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-06456384 trihydrochloride is the trihydrochloride salt of a highly potent and selective NaV1.7 inhibitor developed for preclinical pharmacology and pain model research. The compound has been profiled in vitro and in vivo and is supplied as a high-purity research sample.

  • Salt form: trihydrochloride.
  • Target: voltage-gated sodium channel NaV1.7.
  • Potency: IC50 = 0.01 nM against NaV1.7.
  • Purity: 99.38% by HPLC (supplier specification).
  • Molecular weight: 829.18 g/mol (trihydrochloride salt).
  • Molecular formula: C35H35Cl3F3N7O3S2.
  • Intended use: preclinical pharmacology and formalin pain model research.
  • Supplied quantity: 50 mg research sample.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.