Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-06456384 trihydrochloride | 1834610-75-1 | MFCD31697702 | 99.4% | 829.18 | C35H35Cl3F3N7O3S2 | 10 MG
SDP

Catalog No. 5000228679
Encompass_Preferred

PF-06456384 trihydrochloride is a research-grade small-molecule inhibitor selective for the voltage-gated sodium channel NaV1.7. It is reported to have extremely high potency (IC50 = 0.01 nM) and is supplied at high purity for use in in vitro and in vivo pain-model research.

  • Highly potent NaV1.7 inhibitor (IC50 = 0.01 nM).
  • High purity suitable for research applications (>99%).
  • Molecular weight 829.18 and formula C35H35Cl3F3N7O3S2.
  • Provided as the trihydrochloride salt for enhanced stability and handling.
  • Intended for laboratory research use only.

Catalog No. 50-002-28679 Supplier Medchemexpress LLC Supplier No. HY118952A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-06456384 trihydrochloride is a research-grade small-molecule inhibitor selective for the voltage-gated sodium channel NaV1.7. It is reported to have extremely high potency (IC50 = 0.01 nM) and is supplied at high purity for use in in vitro and in vivo pain-model research.

  • Highly potent NaV1.7 inhibitor (IC50 = 0.01 nM).
  • High purity suitable for research applications (>99%).
  • Molecular weight 829.18 and formula C35H35Cl3F3N7O3S2.
  • Provided as the trihydrochloride salt for enhanced stability and handling.
  • Intended for laboratory research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.