Learn More
Medchemexpress LLC HY-19985 5mg Medchemexpress, PF-06459988 CAS:1428774-45-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY199855MG
Encompass_Preferred
Medchemexpress, HY-19985 5mg PF-06459988 CAS:1428774-45-1 PF-06459988 is an irreversible inhibitor of T790M-Containing EGFR Mutants. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2208