Learn More
Medchemexpress LLC HY-108339 5mg Medchemexpress, PF-06471553 CAS:1808094-07-6 Purity:>98%
SDP
Catalog No. 501968533
Encompass_Preferred
Medchemexpress, HY-108339 5mg PF-06471553 CAS:1808094-07-6 PF-06471553 is a potent, selective and orally available monoacylglycerol acyltransferase 3 (MGAT3) inhibitor, with an IC50 of 92 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8533 Supplier Medchemexpress LLC Supplier No. HY1083395MG
Medchemexpress, HY-108339 5mg PF-06471553 CAS:1808094-07-6 PF-06471553 is a potent, selective and orally available monoacylglycerol acyltransferase 3 (MGAT3) inhibitor, with an IC50 of 92 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.