Learn More
Medchemexpress LLC HY-120789 1mg , PF-06648671 CAS:1587727-31-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1207891MG
Encompass_Preferred
HY-120789 1mg PF-06648671 CAS: 1587727-31-8 PF-06648671 is a novel, brain‐penetrable, and orally active γ‐secretase modulator (GSM). PF-06648671 reduces Aβ42 and Aβ40, with concomitant increases in Aβ37 and Aβ38 in vitro. PF-06648671 is used for the study of Alzheimer’s disease.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1693