Learn More
Cayman Chemical PF-06649751 50mg
SDP
Catalog No. 502950971
Encompass_Preferred
A dopamine D1 and D5 receptor partial agonist; increases locomotor activity and reduces a global parkinsonian disability score in a macaque model of Parkinson's disease induced by MPTP at 0.04 mg/kg
Catalog No. 50-295-0971 Supplier Cayman Chemical Supplier No. 3355650 MG
A dopamine D1 and D5 receptor partial agonist; increases locomotor activity and reduces a global parkinsonian disability score in a macaque model of Parkinson's disease induced by MPTP at 0.04 mg/kg