Learn More
Selleck Chemical LLC PF-06650833 5mg
SDP
Supplier:  Selleck Chemical LLC S85315MG
Encompass_Preferred
PF-06650833 is a potent, selective inhibitor of IRAK4 with an IC50 of 0.2 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3367