Learn More
Selleck Chemical LLC PF-06651600 10mg 1792180-81-4
SDP
Catalog No. 501948186
Encompass_Preferred
PF-06651600 is a potent and irreversible JAK3-selective inhibitor with an IC50 of 33.1 nM but without activity (IC50 > 10 000 nM) against JAK1, JAK2, and TYK2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8186 Supplier Selleck Chemical LLC Supplier No. S853810MG
PF-06651600 is a potent and irreversible JAK3-selective inhibitor with an IC50 of 33.1 nM but without activity (IC50 > 10 000 nM) against JAK1, JAK2, and TYK2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.