Learn More
Medchemexpress LLC HY-103682 5mg Medchemexpress, PF-06726304 CAS:1616287-82-1 Purity:>98%
SDP
Catalog No. 501968160
Encompass_Preferred
Medchemexpress, HY-103682 5mg PF-06726304 CAS:1616287-82-1 PF-06726304 is a potent and selective EZH2 inhibitor with a Ki of 0.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8160 Supplier Medchemexpress LLC Supplier No. HY1036825MG
Medchemexpress, HY-103682 5mg PF-06726304 CAS:1616287-82-1 PF-06726304 is a potent and selective EZH2 inhibitor with a Ki of 0.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.