Learn More
Medchemexpress LLC HY-112706 5mg Medchemexpress, PF-06747711 CAS:1892576-58-7 Purity:>98%
SDP
Catalog No. 501932432
Encompass_Preferred
Medchemexpress, HY-112706 5mg PF-06747711 CAS:1892576-58-7 PF-06747711 is a potent, selective, and orally active retinoic acid receptor-related orphan C2 ( RORC2 , also known as RORγt ) inverse agonist, with an IC 50 of 4.1 nM. Anti-skin inflammatory activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2432 Supplier Medchemexpress LLC Supplier No. HY1127065MG
Medchemexpress, HY-112706 5mg PF-06747711 CAS:1892576-58-7 PF-06747711 is a potent, selective, and orally active retinoic acid receptor-related orphan C2 ( RORC2 , also known as RORγt ) inverse agonist, with an IC 50 of 4.1 nM. Anti-skin inflammatory activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.