Learn More
Medchemexpress LLC HY-112157 10mg Medchemexpress, PF-06751979 CAS:1818339-66-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11215710MG
Encompass_Preferred
Medchemexpress, HY-112157 10mg PF-06751979 CAS:1818339-66-0 PF-06751979 is a potent, brain penetrant, β-site amyloid precursor protein cleaving enzyme 1 (BACE1) inhibitor with an IC50 of 7.3 nM in BACE1 binding assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9043