Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-06815345 (hydrochloride) | 2334434-49-8 | 98.2% | 634.49 | 50 MG
SDP

Catalog No. 5000437624
Encompass_Preferred

PF-06815345 hydrochloride is an orally active and potent inhibitor of proprotein convertase subtilisin/kexin type 9 (PCSK9) with an IC50 value of 13.4 μM. It significantly decreases the PCSK9 level in vivo in mice. This solid chemical is intended for use as a laboratory chemical and in the manufacture of substances. It is for research use only and should be handled by suitably qualified and experienced personnel.

  • Orally active and potent PCSK9 inhibitor
  • Has an IC50 value of 13.4 μM
  • Significantly decreases PCSK9 level in vivo in mice
  • Solid chemical form
  • Molecular formula: C27H30Cl2FN9O4
  • Molecular weight: 634.49
  • For research use only
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-37624 Supplier Medchemexpress LLC Supplier No. HY112598A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-06815345 hydrochloride is an orally active and potent inhibitor of proprotein convertase subtilisin/kexin type 9 (PCSK9) with an IC50 value of 13.4 μM. It significantly decreases the PCSK9 level in vivo in mice. This solid chemical is intended for use as a laboratory chemical and in the manufacture of substances. It is for research use only and should be handled by suitably qualified and experienced personnel.

  • Orally active and potent PCSK9 inhibitor
  • Has an IC50 value of 13.4 μM
  • Significantly decreases PCSK9 level in vivo in mice
  • Solid chemical form
  • Molecular formula: C27H30Cl2FN9O4
  • Molecular weight: 634.49
  • For research use only
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.