Learn More
Medchemexpress LLC PF-06815345 hydrochloride | 2334434-49-8 | 98.2% | 634.49 g/mol | C27H30Cl2FN9O4 | 5 MG
SDP
Catalog No. 5000264324
Encompass_Preferred
PF-06815345 hydrochloride is a small-molecule, orally active inhibitor of proprotein convertase subtilisin/kexin type 9 (PCSK9) provided for research use. It shows in vitro PCSK9 inhibition (IC50 13.4 μM) and reduces PCSK9 levels in vivo in mice. The compound is supplied as the hydrochloride salt, formula C27H30Cl2FN9O4, molecular weight 634.49 g/mol.
Catalog No. 50-002-64324 Supplier Medchemexpress LLC Supplier No. HY112598A5MG
PF-06815345 hydrochloride is a small-molecule, orally active inhibitor of proprotein convertase subtilisin/kexin type 9 (PCSK9) provided for research use. It shows in vitro PCSK9 inhibition (IC50 13.4 μM) and reduces PCSK9 levels in vivo in mice. The compound is supplied as the hydrochloride salt, formula C27H30Cl2FN9O4, molecular weight 634.49 g/mol.