Learn More
Medchemexpress LLC HY-101111 100mg Medchemexpress, PF-06840003 CAS:198474-05-4 Purity:>98%
SDP
Catalog No. 501932045
Encompass_Preferred
Medchemexpress, HY-101111 100mg PF-06840003 CAS:198474-05-4 PF-06840003 is a highly selective orally bioavailable IDO-1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2045 Supplier Medchemexpress LLC Supplier No. HY101111100MG
Medchemexpress, HY-101111 100mg PF-06840003 CAS:198474-05-4 PF-06840003 is a highly selective orally bioavailable IDO-1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.