Learn More
TARGETMOL CHEMICALS INC PF-06873600 5MG
SDP
Catalog No. 502579071
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-06873600 is a selective and orally bioavailable cyclin-dependent kinase (CDK) inhibitor(CDK2 CDK4 and CDK6 with Ki of 0.09 nM 0.13 nM and 0.16 nM respectively) has potential antineoplastic activity. purity: 99%
Catalog No. 50-257-9071 Supplier TARGETMOL CHEMICALS INC Supplier No. T84635MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-06873600 is a selective and orally bioavailable cyclin-dependent kinase (CDK) inhibitor(CDK2 CDK4 and CDK6 with Ki of 0.09 nM 0.13 nM and 0.16 nM respectively) has potential antineoplastic activity. purity: 99%