Learn More
Selleck Chemical LLC PF-06873600 S8816-5MG
SDP
Catalog No. 5000024738
Encompass_Preferred
PF-06873600 is an inhibitor of cyclin-dependent kinase 2 (CDK2) CDK4 and CDK6 with Ki values of 0 1 1 2 and 0 1 nM respectively blocks the phosphorylation of retinoblastoma protein (RB1) and limits the proliferation of OVCAR-3 ovarian cancer cells with EC50s of 19 and 45 nM respectively
Catalog No. 50-000-24738 Supplier Selleck Chemical LLC Supplier No. S88165MG
PF-06873600 is an inhibitor of cyclin-dependent kinase 2 (CDK2) CDK4 and CDK6 with Ki values of 0 1 1 2 and 0 1 nM respectively blocks the phosphorylation of retinoblastoma protein (RB1) and limits the proliferation of OVCAR-3 ovarian cancer cells with EC50s of 19 and 45 nM respectively