Learn More
Medchemexpress LLC PF-06928215 10mg | 2378173-15-8 | 10 MG
SDP
Catalog No. 5000226479
Encompass_Preferred
PF-06928215 is a small-molecule inhibitor of cyclic GMP-AMP synthase (cGAS) with a reported IC50 of 4.9 μM and a binding affinity (Kd) of 0.2 μM. It is supplied for research use as a high-purity solid with formula C20H20N4O4 and molecular weight 380.40 g/mol.
  • Inhibits cGAS with reported IC50 of 4.9 μM.
  • High binding affinity (Kd 0.2 μM).
  • Chemical formula C20H20N4O4 and molecular weight 380.40 g/mol.
  • High purity (99.7%) suitable for research use.
  • Provided as a solid; typical pack size 10 MG.
Catalog No. 50-002-26479 Supplier Medchemexpress LLC Supplier No. HY11418210MG
PF-06928215 is a small-molecule inhibitor of cyclic GMP-AMP synthase (cGAS) with a reported IC50 of 4.9 μM and a binding affinity (Kd) of 0.2 μM. It is supplied for research use as a high-purity solid with formula C20H20N4O4 and molecular weight 380.40 g/mol.
  • Inhibits cGAS with reported IC50 of 4.9 μM.
  • High binding affinity (Kd 0.2 μM).
  • Chemical formula C20H20N4O4 and molecular weight 380.40 g/mol.
  • High purity (99.7%) suitable for research use.
  • Provided as a solid; typical pack size 10 MG.