Learn More
Cayman Chemical PF-06928215, Purity: ≥98%, CAS: 2378173-15-8, 1 mg
SDP
Catalog No. 502946593
Encompass_Preferred
A cGAS inhibitor (IC50 = 4.9 µM in a fluorescence polarization assay)
Catalog No. 50-294-6593 Supplier Cayman Chemical Supplier No. 325151 MG
A cGAS inhibitor (IC50 = 4.9 µM in a fluorescence polarization assay)