Learn More
Medchemexpress LLC HY-136244 5mg Medchemexpress, PF-06952229 CAS:1801333-55-0 Purity:>98%
SDP
Catalog No. 502030409
Encompass_Preferred
Medchemexpress, HY-136244 5mg PF-06952229 CAS:1801333-55-0 PF-06952229 is a potent, selective and orally active TGFbR1 inhibitor. PF-06952229 specifically binds to TGFbR1 and prevents TGFbR1-mediated signal transduction. PF-06952229 is a promising antineoplastic agent for the study solid tumors, especifically metastatic breast cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0409 Supplier Medchemexpress LLC Supplier No. HY1362445MG
Medchemexpress, HY-136244 5mg PF-06952229 CAS:1801333-55-0 PF-06952229 is a potent, selective and orally active TGFbR1 inhibitor. PF-06952229 specifically binds to TGFbR1 and prevents TGFbR1-mediated signal transduction. PF-06952229 is a promising antineoplastic agent for the study solid tumors, especifically metastatic breast cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.