Learn More
Medchemexpress LLC Pf-07038124 hydrochloride | 2415317-57-4 | 99.3% | 363.64 g/mol | C18H23BClNO4 | 5 MG
SDP
Catalog No. 5000221629
Encompass_Preferred
PF-07038124 hydrochloride is the hydrochloride salt of a selective phosphodiesterase 4 (PDE4) inhibitor supplied for laboratory research into inflammatory skin diseases and immune signaling pathways. Intended for research use only; not for human or clinical use.
  • High purity suitable for biochemical and cellular assays.
  • Potent inhibition of PDE4B2 (IC50 ≈ 0.5 nM).
  • Demonstrated cytokine inhibition of IL-13, IL-4, and IFN-γ in reported assays.
  • Available in small milligram quantities for early-stage studies.
  • Hydrochloride salt form improves solubility for aqueous assays.
Catalog No. 50-002-21629 Supplier Medchemexpress LLC Supplier No. HY144683A5MG
PF-07038124 hydrochloride is the hydrochloride salt of a selective phosphodiesterase 4 (PDE4) inhibitor supplied for laboratory research into inflammatory skin diseases and immune signaling pathways. Intended for research use only; not for human or clinical use.
  • High purity suitable for biochemical and cellular assays.
  • Potent inhibition of PDE4B2 (IC50 ≈ 0.5 nM).
  • Demonstrated cytokine inhibition of IL-13, IL-4, and IFN-γ in reported assays.
  • Available in small milligram quantities for early-stage studies.
  • Hydrochloride salt form improves solubility for aqueous assays.