Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-07038124 hydrochloride | 2415317-57-4 | 99.3% | C18H23BClNO4 | 1 MG
SDP

Catalog No. 5000355877
Encompass_Preferred

PF-07038124 hydrochloride is a potent PDE4 inhibitor with an IC50 of 0.5 nM for PDE4B2. It demonstrates immunomodulatory effects by inhibiting IL-13, IL4, and IFNγ, making it suitable for research related to atopic dermatitis and plaque psoriasis. This compound has been investigated in Phase 1 and Phase 2 clinical trials for these conditions. It is supplied as an off-white to light yellow solid.

  • Specifically targets PDE4B2 with high potency
  • Shows inhibitory activity against IL-13, IL4, and IFNγ
  • Used in the study of atopic dermatitis and plaque psoriasis
  • Investigated in Phase 1 and Phase 2 clinical trials
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage is 4°C sealed, away from moisture

Catalog No. 50-003-55877 Supplier Medchemexpress LLC Supplier No. HY144683A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-07038124 hydrochloride is a potent PDE4 inhibitor with an IC50 of 0.5 nM for PDE4B2. It demonstrates immunomodulatory effects by inhibiting IL-13, IL4, and IFNγ, making it suitable for research related to atopic dermatitis and plaque psoriasis. This compound has been investigated in Phase 1 and Phase 2 clinical trials for these conditions. It is supplied as an off-white to light yellow solid.

  • Specifically targets PDE4B2 with high potency
  • Shows inhibitory activity against IL-13, IL4, and IFNγ
  • Used in the study of atopic dermatitis and plaque psoriasis
  • Investigated in Phase 1 and Phase 2 clinical trials
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage is 4°C sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.