Learn More
Ubpbio A covalent inhibitor of 3CL protease. A component of Paxlovid used for the treatment of COVID 19. CAS# 26288280-40-8, > 99% purity
Catalog No. 502339305
Encompass_Preferred
PF-07321332 is an orally active covalent inhibitor of the 3C-like protease by direct binding of the catalytic cysteine 145. 3CL protease cleaves polyproteins 1a and 1ab of SARS-CoV-2, a step required for reproduce the virus. The PF-07321332/ritonavir combination drug (trade name Paxlovid) is efficient for the treatment of COVID-19 as, in November 2021, Pfizer announced phase 2/3 clinal trial results that the drug treatment reduced 89% in hospitalizations when given within three days after symptom onset. This product is for research use only.
Catalog No. 50-233-9305 Supplier Ubpbio Supplier No. F100010
PF-07321332 is an orally active covalent inhibitor of the 3C-like protease by direct binding of the catalytic cysteine 145. 3CL protease cleaves polyproteins 1a and 1ab of SARS-CoV-2, a step required for reproduce the virus. The PF-07321332/ritonavir combination drug (trade name Paxlovid) is efficient for the treatment of COVID-19 as, in November 2021, Pfizer announced phase 2/3 clinal trial results that the drug treatment reduced 89% in hospitalizations when given within three days after symptom onset. This product is for research use only.