Learn More
Selleck Chemical LLC PF-07799933 E4681-5MG
SDP
Catalog No. 5000039452
Encompass_Preferred
PF-07799933 (Claturafenib ARRY-440) is a brain-penetrant selective pan-mutant inhibitor of BRAF It exhibits broad inhibition of pERK levels in HT29 cells with an IC50 value of 1 6 nM PF-07799933 treatment displays antitumor activity against BRAFV600- and non-V600-mutant cancers preclinically and in treatment-refractory patients
Catalog No. 50-000-39452 Supplier Selleck Chemical LLC Supplier No. E46815MG
PF-07799933 (Claturafenib ARRY-440) is a brain-penetrant selective pan-mutant inhibitor of BRAF It exhibits broad inhibition of pERK levels in HT29 cells with an IC50 value of 1 6 nM PF-07799933 treatment displays antitumor activity against BRAFV600- and non-V600-mutant cancers preclinically and in treatment-refractory patients