Learn More
Medchemexpress LLC HY-12361 5mg Medchemexpress, PF 1022A CAS:133413-70-4 Purity:>98%
SDP
Catalog No. 502081021
Encompass_Preferred
Medchemexpress, HY-12361 5mg PF 1022A CAS:133413-70-4 PF 1022A is a cyclooctadepsipeptide with broadspectrum anthelmintic properties produced by fermentation of the fungus Mycelia sterilia. PF 1022A is a channel-forming ionophore. PF 1022A showes strong anthelmintic activities against Ascaridia galli in chickens. PF 1022A also can be used for angiostrongyliasis research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1021 Supplier Medchemexpress LLC Supplier No. HY123615MG
Medchemexpress, HY-12361 5mg PF 1022A CAS:133413-70-4 PF 1022A is a cyclooctadepsipeptide with broadspectrum anthelmintic properties produced by fermentation of the fungus Mycelia sterilia. PF 1022A is a channel-forming ionophore. PF 1022A showes strong anthelmintic activities against Ascaridia galli in chickens. PF 1022A also can be used for angiostrongyliasis research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.