Learn More
Medchemexpress LLC HY-100873 5mg Medchemexpress, PF-1355 CAS:1435467-38-1 Purity:>98%
SDP
Catalog No. 501967786
Encompass_Preferred
Medchemexpress, HY-100873 5mg PF-1355 CAS:1435467-38-1 PF-1355 is a selective 2-thiouracil mechanism-based MPO inhibitor, used for treatment of vasculitic diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7786 Supplier Medchemexpress LLC Supplier No. HY1008735MG
Medchemexpress, HY-100873 5mg PF-1355 CAS:1435467-38-1 PF-1355 is a selective 2-thiouracil mechanism-based MPO inhibitor, used for treatment of vasculitic diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.