Learn More
Enzo Life Sciences PF-356231 (5mg). CAS: 766536-21-4
SDP
Catalog No. 502011493
Encompass_Preferred
Specific non-chelating small-molecule inhibitor of MMP-12 (IC50 = 14 nM). At higher concentrations it inhibits other MMPs, but spares MMP-1, making it a valuable tool for investigation of MMP activity and function. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1493 Supplier Enzo Life Sciences Supplier No. BMLPI1550005
Specific non-chelating small-molecule inhibitor of MMP-12 (IC50 = 14 nM). At higher concentrations it inhibits other MMPs, but spares MMP-1, making it a valuable tool for investigation of MMP activity and function. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.