Learn More
Medchemexpress LLC HY-107427 5mg Medchemexpress, PF-3644022 CAS:1276121-88-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1074275MG
Encompass_Preferred
Medchemexpress, HY-107427 5mg PF-3644022 CAS:1276121-88-0 PF-3644022 is a potent, selective, orally active and ATP-competitive MAPKAPK2 (MK2) inhibitor with an IC50 of 5.2 nM and a Ki of 3 nM. PF-3644022 also inhibits MK3 and p38 regulated/activated kinase (PRAK) with IC50s of 53 nM and 5.0 nM, respectively. PF-3644022 potently inhibits TNFα production and has anti-inflammatory effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8413