Learn More
Selleck Chemical LLC PF-3758309 10mg 898044-15-0 PF-03758309
SDP
Supplier:  Selleck Chemical LLC S709410MG
Encompass_Preferred
PF-03758309 (PF-03758309) is a potent, ATP-competitive, pyrrolopyrazole inhibitor of PAK4 with IC50 of 1.3 nM. PF-3758309 is antiproliferative and induces apoptosis in a HCT116 tumor model. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0951591