Learn More
Medchemexpress LLC HY-13007 10mg Medchemexpress, PF-3758309 CAS:898044-15-0 Purity:>98%
SDP
Catalog No. 502028938
Encompass_Preferred
Medchemexpress, HY-13007 10mg PF-3758309 CAS:898044-15-0 PF-3758309 is a potent, orally available, and reversible ATP-competitive inhibitor of PAK4 (Kd= 2.7 nM; Ki=18.7 nM). PF-3758309 has the expected cellular functions of a PAK4 inhibitor: inhibition of anchorage-independent growth, induction of apoptosis, cytoskeletal remodeling, and inhibition of proliferation[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8938 Supplier Medchemexpress LLC Supplier No. HY1300710MG
Medchemexpress, HY-13007 10mg PF-3758309 CAS:898044-15-0 PF-3758309 is a potent, orally available, and reversible ATP-competitive inhibitor of PAK4 (Kd= 2.7 nM; Ki=18.7 nM). PF-3758309 has the expected cellular functions of a PAK4 inhibitor: inhibition of anchorage-independent growth, induction of apoptosis, cytoskeletal remodeling, and inhibition of proliferation[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.