Learn More
Apexbio Technology LLC PF-3758309 898044-15-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A37165.1
Encompass_Preferred
PF-3758309 (CAS 898044-15-0) is a selective ATP-competitive inhibitor of p21-activated kinase 4 (PAK4) a serine/threonine kinase involved in cytoskeletal reorganization and cellular signaling mediated by Cdc42-GTP and JNK-GTP interactions PF-3758309 inhibits phosphorylation of PAK4 substrates (e g GEF-H1 IC50 1 3 nM) and suppresses PAK4-mediated anchorage-independent growth In xenograft tumor models (HCT116 A549) oral administration of PF-3758309 significantly reduced tumor growth demonstrating its utility in research investigating PAK-dependent oncogenic pathways
Catalog No. 50-000-64016