Learn More
Apexbio Technology LLC PF-3758309, 50mg. Cas: 898044-15-0 MFCD: MFCD18633226. PAK4 inhibitor
SDP
Supplier:  Apexbio Technology LLC A371650
Encompass_Preferred
PF-3758309 is an inhbitor of PAK4 with IC50 of 1.3 nM [1].P21-activated kinases (PAKs) are the family of serine/theronine kinases, which play important role in linking Rho GTPase to cytoskeleton reorganization and nuclear signaling. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4828