Learn More
TARGETMOL CHEMICALS INC PF-3758309 HYDROCHLORIDE 10MG
SDP
Catalog No. 502576786
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-3758309 hydrochloride (PF-03758309 hydrochloride) is a PAK4 inhibitor is also a n orally bioavailable small-molecule inhibitor of p21-activated kinase 4 (PAK4) with potential antineoplastic activity. PF-3758309 hydrochloride binds to PAK4 inhibiting PAK4 activity and cancer cell growth. PAK4 a serine/threonine kinase belonging to the p21-activated kinase (PAK) family is often upregulated in a variety of cancer cell types and plays an important role in cancer cell motility proliferation and survival. purity: 98%
Catalog No. 50-257-6786 Supplier TARGETMOL CHEMICALS INC Supplier No. T423010MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-3758309 hydrochloride (PF-03758309 hydrochloride) is a PAK4 inhibitor is also a n orally bioavailable small-molecule inhibitor of p21-activated kinase 4 (PAK4) with potential antineoplastic activity. PF-3758309 hydrochloride binds to PAK4 inhibiting PAK4 activity and cancer cell growth. PAK4 a serine/threonine kinase belonging to the p21-activated kinase (PAK) family is often upregulated in a variety of cancer cell types and plays an important role in cancer cell motility proliferation and survival. purity: 98%