Learn More
TARGETMOL CHEMICALS INC PF-3845 25MG
SDP
Catalog No. 502579620
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-3845 is a potent selective and irreversible FAAH inhibitor with Ki of 230 nM showing negligible activity against FAAH2. purity: 99%
Catalog No. 50-257-9620 Supplier TARGETMOL CHEMICALS INC Supplier No. T604325MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-3845 is a potent selective and irreversible FAAH inhibitor with Ki of 230 nM showing negligible activity against FAAH2. purity: 99%