Learn More
Medchemexpress LLC N-3-pyridinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methyl]-1-piperidinecarboxamide | 1196109-52-0 | MFCD21365072 | >98.0% | 456.46 g/mol | C24H23F3N4O2 | 5 MG
SDP
Catalog No. 502081595
Encompass_Preferred
PF-3845 is a selective, irreversible fatty acid amide hydrolase (FAAH) inhibitor for research use. It covalently carbamylates the catalytic serine of FAAH, elevating endocannabinoid levels and producing anti-inflammatory and analgesic effects in preclinical studies. Supplied as a white to off-white solid suitable for biochemical and in vivo experiments.
  • Selective, irreversible FAAH inhibition.
  • Carbamylates the catalytic serine of FAAH.
  • Demonstrated anti-inflammatory and analgesic effects in preclinical models.
  • Available as a high-purity solid (>98%).
  • Suitable for biochemical and in vivo neuroscience research.
Catalog No. 50-208-1595 Supplier Medchemexpress LLC Supplier No. HY143805MG
PF-3845 is a selective, irreversible fatty acid amide hydrolase (FAAH) inhibitor for research use. It covalently carbamylates the catalytic serine of FAAH, elevating endocannabinoid levels and producing anti-inflammatory and analgesic effects in preclinical studies. Supplied as a white to off-white solid suitable for biochemical and in vivo experiments.
  • Selective, irreversible FAAH inhibition.
  • Carbamylates the catalytic serine of FAAH.
  • Demonstrated anti-inflammatory and analgesic effects in preclinical models.
  • Available as a high-purity solid (>98%).
  • Suitable for biochemical and in vivo neuroscience research.