Learn More
Abcam PF-3845, FAAH inhibitor, 25MG
Catalog No. 502707158
Encompass_Preferred
MW 456.47 g/mol, Purity >98%. Selective, irreversible FAAH inhibitor (Ki = 0.23 μM). Anti-inflammatory effects in vivo. Crosses the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7158 Supplier Abcam Supplier No. AB12089425MG
MW 456.47 g/mol, Purity >98%. Selective, irreversible FAAH inhibitor (Ki = 0.23 μM). Anti-inflammatory effects in vivo. Crosses the blood-brain barrier.
The product is subject to the following: Abcam Restricted Use Statement