Learn More
VAI PF-4 Sterile Face Mask
Catalog No. 19546530 Shop All VAI Products
Change view
Click to view available options
PF-4 Sterile Face Mask; Fastener: 2 Elastic Straps
Catalog No.
19-546-530 PF-4 Sterile Face Mask; Fastener: 2 Elastic Straps
1 options
This item is not returnable. View return policy
Catalog No. 19-546-530 Supplier VAI Supplier No. PF4SM2S2029
Add to Cart
Add to Cart
This item is not returnable. View return policy
PF-4 Sterile Face Mask is constructed of 3 layers of non-woven material with two silicon-free elastic headloop bands.
  • Material is hypoallergenic and soft to prevent skin irritation
  • Coated metal strip acts as a nose piece to keep the mask secure
  • Intended uses include sterile manufacturing and for the prevention of small particle inhalation
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Elastic Straps
Size Standard
Sterility Sterile
Construction 3-ply
Disposable Single-use
Model PF-4SM-2-S-2029
Packaging Quantity 1 Ea.
Packaging Type Individually Bagged
Series PF-4