Learn More
Medchemexpress LLC HY-13245 10mg , PF-4136309 CAS:1341224-83-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1324510MG
Encompass_Preferred
HY-13245 10mg PF-4136309 CAS: 1341224-83-6 PF-4136309 is a potent, selective, and orally bioavailable CCR2 antagonist, with IC50s of 5.2 nM, 17 nM and 13 nM for human, mouse and rat CCR2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9690