Learn More
Medchemexpress LLC HY-13245 50mg , PF-4136309 INCB8761 CAS:1341224-83-6 Purity:98%
SDP
Catalog No. 501493877
Encompass_Preferred
Medchemexpress, HY-13245 50mg PF-4136309 INCB8761 CAS:1341224-83-6 Formula:C29H31F3N6O3 IC50: 5.2 nM (Human CCR2), 17 nM (Mouse CCR2), 13 nM (Rat CCR2) Purity:98% PF-4136309 is a potent, selective, and orally bioavailable CCR2 antagonist, with IC 50 of 5.2 nM, 17 nM and 13 nM for human, mouse and rat CCR2. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3877 Supplier Medchemexpress LLC Supplier No. HY1324550MG
Medchemexpress, HY-13245 50mg PF-4136309 INCB8761 CAS:1341224-83-6 Formula:C29H31F3N6O3 IC50: 5.2 nM (Human CCR2), 17 nM (Mouse CCR2), 13 nM (Rat CCR2) Purity:98% PF-4136309 is a potent, selective, and orally bioavailable CCR2 antagonist, with IC 50 of 5.2 nM, 17 nM and 13 nM for human, mouse and rat CCR2. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.