Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PF-4191834 100mg | 1029317-21-2 | 100 MG
SDP

Catalog No. 5000307651
Encompass_Preferred

PF-4191834 is an orally active, non-iron-chelating, non-redox inhibitor of 5-lipoxygenase (5-LOX) with reported IC50 = 229 nM and high selectivity versus 12-LOX and 15-LOX. It is used in enzyme, cell-based, and in vivo inflammation models for research on 5-LOX-mediated pathways.

  • Orally active 5-LOX inhibitor (IC50 = 229 nM).
  • Approximately 300-fold selectivity over 12-LOX and 15-LOX.
  • High purity (99.77%).
  • Available as solid; common package 100 MG.
  • Molecular weight 393.5 g/mol; formula C22H23N3O2S.
  • Recommended storage: powder at -20°C for long-term stability.

Catalog No. 50-003-07651 Supplier Medchemexpress LLC Supplier No. HY117048100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-4191834 is an orally active, non-iron-chelating, non-redox inhibitor of 5-lipoxygenase (5-LOX) with reported IC50 = 229 nM and high selectivity versus 12-LOX and 15-LOX. It is used in enzyme, cell-based, and in vivo inflammation models for research on 5-LOX-mediated pathways.

  • Orally active 5-LOX inhibitor (IC50 = 229 nM).
  • Approximately 300-fold selectivity over 12-LOX and 15-LOX.
  • High purity (99.77%).
  • Available as solid; common package 100 MG.
  • Molecular weight 393.5 g/mol; formula C22H23N3O2S.
  • Recommended storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.