Learn More
Medchemexpress LLC PF-4191834 25mg | 1029317-21-2 | 393.50 g·mol⁻1 | C22H23N3O2S | 25 MG
SDP
Catalog No. 5000308347
Encompass_Preferred
PF-4191834 is a research-grade small molecule 5-lipoxygenase (5-LOX) inhibitor for preclinical and in vitro studies. It is described as orally active, non-iron chelating, and non-redox, provided as a high-purity solid suitable for biochemical assays and cellular pharmacology research.
  • 5-lipoxygenase inhibitor for biochemical and cellular assays.
  • Orally active small molecule suitable for preclinical studies.
  • Non-iron chelating and non-redox mechanism of action.
  • High purity solid (manufacturer lists 99.77% purity).
  • Molecular weight 393.50 g·mol⁻1; formula C22H23N3O2S.
  • Available in small sample sizes for laboratory research.
Catalog No. 50-003-08347 Supplier Medchemexpress LLC Supplier No. HY11704825MG
PF-4191834 is a research-grade small molecule 5-lipoxygenase (5-LOX) inhibitor for preclinical and in vitro studies. It is described as orally active, non-iron chelating, and non-redox, provided as a high-purity solid suitable for biochemical assays and cellular pharmacology research.
  • 5-lipoxygenase inhibitor for biochemical and cellular assays.
  • Orally active small molecule suitable for preclinical studies.
  • Non-iron chelating and non-redox mechanism of action.
  • High purity solid (manufacturer lists 99.77% purity).
  • Molecular weight 393.50 g·mol⁻1; formula C22H23N3O2S.
  • Available in small sample sizes for laboratory research.